missing translation for 'onlineSavingsMsg'
Learn More

Sulfadoxine, MedChemExpress

Product Code. 16480928
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480928 5 g 5g
16478488 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480928 Supplier Medchem Express Supplier No. HYB0439
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulfadoxine(Sulphadoxine) is a long acting sulfonamide that is used, usually in combination with other drugs, for respiratory, urinary tract and malarial infections. Sulfadoxine inhibits HIV replication in peripheral blood mononuclear cells.

Sulfadoxine(Sulphadoxine) is a long acting sulfonamide that is used, usually in combination with other drugs, for respiratory, urinary tract and malarial infections. Sulfadoxine inhibits HIV replication in peripheral blood mononuclear cells.

Specifications

Chemical Name or Material Sulfadoxine
Physical Form Solid
CAS 2447-57-6
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C12H14N4O4S
For Use With (Application) COVID-19-anti-virus
Synonym Sulphadoxine
Solubility Information DMSO : ≥ 100 mg/mL (322.24 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=S(NC1=NC=NC(OC)=C1OC)(C2=CC=C(N)C=C2)=O
Molecular Weight (g/mol) 310.33
Formula Weight 310.33
Percent Purity 99.44%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.