missing translation for 'onlineSavingsMsg'
Learn More

Sulfadoxin 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30313452
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313452 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313452 Supplier Dr. Ehrenstorfer Supplier No. DREA16990600AL1000
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfadoxine
CAS 2447-57-6
Analyte or Component Names Sulfadoxine
Shipping Condition +20°C
Molecular Formula C12 H14 N4 O4 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Sulfadoxine, 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(5,6-dimethoxy-4-pyrimidinyl)-, 4,5-Dimethoxy-6-sulfanilamidopyrimidine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzensulfonamide, 4-Sulfanilamido-5,6-dimethoxypyrimidine, Fanasil, Fanasulf, N1-5,6-Dimethoxy-4-pyrimidinylsulfanilamide, Orthosulfin, Ro 4-4393, Sanasil, Sulfadoxin, Sulformethoxine, Sulformetoxin, Sulformetoxine, Sulforthodimethoxine, Sulforthomidine, WR 4073
InChI Formula InChI=1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)
SMILES COc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC
IUPAC Name 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 310.33
Quantity 1 mL
Formula Weight 310.0736 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.