missing translation for 'onlineSavingsMsg'
Learn More

Sulfadimethoxine, Thermo Scientific™

Product Code. 17780669 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 17780669

Brand: Thermo Scientific Acros 469760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Sulfadimethoxine
Type Sulfadimethoxine
Physical Form Crystalline Powder
CAS 122-11-2
Melting Point 195°C to 203°C
Infrared Spectrum Conforms
Loss on Drying ≤0.5 % (1 g, 105°C)
Packaging Glass Bottle
Color Cream to White
Quantity 5 g
Molecular Formula C12 H14 N4 O4 S
Product Line Life science
MDL Number 00057345
Shipping Condition No special requirements
Molecular Weight (g/mol) 310.33
Percent Purity ≥97.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.