missing translation for 'onlineSavingsMsg'
Learn More

Sulfadimethoxine-d4, TRC

Product Code. 30418437
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418437 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30418437 Supplier Toronto Research Chemicals Supplier No. TRCS6990611MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfadimethoxine-d4
CAS 1020719-80-5
Purity Grade Notes HPLC
Molecular Formula C12H10D4N4O4S
Quantity 1 mg
Synonym Sulfadimethoxine D4
InChI Formula InChI=1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)/i3D,4D,5D,6D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2cc(OC)nc(OC)n2
Molecular Weight (g/mol) 314.35
Formula Weight 314.0987
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.