missing translation for 'onlineSavingsMsg'
Learn More

Sulfaclozine, MedChemExpress

Product Code. 16417586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16417586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417586 Supplier Medchem Express Supplier No. HY19285
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulfaclozine (Sulfachloropyrazine) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. Sulfaclozine is commonly used for the treatment of various poultry diseases (particularly, collibacteriosis, fowl cholera and coccidiosis).

Sulfaclozine (Sulfachloropyrazine) is an efficacious sulphonamide derivative with antibacterial and anticoccidial effects. Sulfaclozine is commonly used for the treatment of various poultry diseases (particularly, collibacteriosis, fowl cholera and coccidiosis).

Specifications

Chemical Name or Material Sulfaclozine
CAS 102-65-8
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H9ClN4O2S
For Use With (Application) COVID-19-immunoregulation
Synonym Sulfachloropyrazine
Solubility Information DMSO : ≥ 29 mg/mL (101.85 mM)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NC(Cl)=CN=C2)=O
Molecular Weight (g/mol) 284.72
Formula Weight 284.72
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.