missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfachloropyridazine, MedChemExpress
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Sulfachloropyridazine is a broad spectrum sulfonamide used against both Gram-positive and Gram-negative aerobic bacteria.
Specifications
Specifications
| Chemical Name or Material | Sulfachloropyridazine |
| CAS | 80-32-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C10H9ClN4O2S |
| Quantity | 250 mg |
| Synonym | Sulfachlorpyridazine |
| Solubility Information | DMSO : 160 mg/mL (561.96 mM; Need ultrasonic and warming) |
| SMILES | O=S(C1=CC=C(N)C=C1)(NC2=NN=C(Cl)C=C2)=O |
| Molecular Weight (g/mol) | 284.72 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction