missing translation for 'onlineSavingsMsg'
Learn More

Sulfacetamide Sodium Salt Monohydrate, TRC

Product Code. 30427546
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427546 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427546 Supplier Toronto Research Chemicals Supplier No. TRCS68896510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfacetamide sodium salt
CAS 6209-17-2
Molecular Formula C8H9N2O3S.H2O.Na
Quantity 10 g
Synonym Sulfacetamide Sodium Monohydrate, N-[(4-Aminophenyl)sulfonyl]acetamide sodium salt hydrate (1:1:1), N-[(4-Aminophenyl)sulfonyl]acetamide monosodium salt monohydrate, N-Sulfanilylacetamide monosodium salt monohydrate, N1-Acetylsulfanilamide sodium monohydrate, Sodium N-sulfanilylacetamide monohydrate, Sodium sulfacetamide monohydrate, Sulamyd sodium monohydrate, Sulfacetamide sodium monohydrate
InChI Formula InChI=1S/C8H10N2O3S.Na.H2O/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;;/h2-5H,9H2,1H3,(H,10,11);;1H2/q;+1;/p-1
SMILES O.[Na+].CC(=O)[N-]S(=O)(=O)c1ccc(N)cc1
IUPAC Name sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate
Molecular Weight (g/mol) 254.2387
Formula Weight 254.0337
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.