missing translation for 'onlineSavingsMsg'
Learn More

Sulfacetamide sodium salt hydrate, 99%

Product Code. 11490557
Click to view available options
Quantity:
25 g
100 g
500 g
Förpackningsstorlek
100g
25g
500g
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 11490557

missing translation for 'mfr': Thermo Scientific Alfa Aesar B22426.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

CAS: 6209-17-2 | C8H9N2NaO3S | 236.22 g/mol

Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfacetamide sodium salt hydrate is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections. It adapts to cure trachoma, conjunctivitis, keratitis.

Solubility
Freely soluble in water. Insoluble in chloroform, ether, and benzene.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 6209-17-2
Molecular Formula C8H9N2NaO3S
Molecular Weight (g/mol) 236.22
MDL Number MFCD00007885
InChI Key PQMSFAORUFMASU-UHFFFAOYSA-M
Synonym sodium sulfacetamide, sulfacetamide sodium monohydrate, sulfacetamide sodium salt hydrate, sulfacetamide sodic hydrate, unii-4nrt660kjq, sulfacetamide sodium usp, cetamide, sulfair forte, sulfair 10, 4nrt660kjq
PubChem CID 6419954
ChEBI CHEBI:63858
IUPAC Name sodium;acetyl-(4-aminophenyl)sulfonylazanide;hydrate
SMILES CC(=O)N([Na])S(=O)(=O)C1=CC=C(N)C=C1

Specifikationer

Quantity 500 g
Solubility Information Freely soluble in water. Insoluble in chloroform,ether,and benzene.
Formula Weight 236.23 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Sulfacetamide sodium salt hydrate

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.