missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfacetamide, Dr. Ehrenstorfer
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Sulfacetamide |
| CAS | 144-80-9 |
| Analyte or Component Names | Sulfacetamide |
| Shipping Condition | +20°C |
| Molecular Formula | C8 H10 N2 O3 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | N-[(4-Aminophenyl)sulfonyl]acetamide, Sulfacetamide, Sulfadimidine Imp. E (Pharmeuropa), Sulfadimidine Impurity E |
| InChI Formula | InChI=1S/C8H10N2O3S/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8/h2-5H,9H2,1H3,(H,10,11) |
| SMILES | CC(=O)NS(=O)(=O)c1ccc(N)cc1 |
| IUPAC Name | N-(4-aminophenyl)sulfonylacetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction