missing translation for 'onlineSavingsMsg'
Learn More

Sulfacetamide, 98%, Thermo Scientific™

Product Code. p-7077287 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 15525106

Brand: Thermo Scientific Acros 442841000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-80-9
Molecular Formula C8H10N2O3S
Molecular Weight (g/mol) 214.239
InChI Key SKIVFJLNDNKQPD-UHFFFAOYSA-N
PubChem CID 5320
ChEBI CHEBI:63845
IUPAC Name N-(4-aminophenyl)sulfonylacetamide
SMILES CC(=O)NS(=O)(=O)C1=CC=C(C=C1)N

Specifications

Type Sulfacetamide
Melting Point 182.0°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C, 2 hrs)
Quantity 100 g
Formula Weight 214.24
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Sulfacetamide
compliance-icons
Product Identifier
  • Sulfacetamide
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.