missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115984
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Sulfatase 1.
Heparan sulfate proteoglycans act as coreceptors for numerous heparin-binding growth factors and cytokines and are involved in cell signaling. Heparan sulfate 6-O-endosulfatases, such as SULF1, selectively remove 6-O-sulfate groups from heparan sulfate. This activity modulates the effects of heparan sulfate by altering binding sites for signaling molecules.
Specifications
| SULF1 | |
| Polyclonal | |
| Unconjugated | |
| SULF1 | |
| AI467640; AW121680; extracellular sulfatase Sulf-1; FLJ30905; FLJ38022; FLJ41750; hSulf-1; Kiaa1077; mKIAA1077; mSulf-1; RSulfFP1; Sulf1; SULF-1; sulfatase 1; sulfatase FP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171396, 23213, 240725 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8IWU6, Q8K007, Q8VI60 | |
| SULF1 | |
| A synthesized peptide derived from human SULF1(Accession Q8IWU6), corresponding to amino acid residues L412-Q462. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction