missing translation for 'onlineSavingsMsg'
Learn More

Sulcotrione, MedChemExpress

Product Code. 16455086
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16455086 100 mg 100mg
16400477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16455086 Supplier Medchem Express Supplier No. HY107368
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulcotrione is a β-triketone herbicide which can inhibit hydroxyphenylpyruvate dioxygenase (HPPD).

Sulcotrione is a β-triketone herbicide which can inhibit hydroxyphenylpyruvate dioxygenase (HPPD).

Specifications

Chemical Name or Material Sulcotrione
CAS 99105-77-8
Color White
Purity Grade Notes Research
Molecular Formula C14H13ClO5S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (304.16 mM; Need ultrasonic and warming)
SMILES O=C(C1C(CCCC1=O)=O)C2=C(C=C(S(C)(=O)=O)C=C2)Cl
Molecular Weight (g/mol) 328.77
Formula Weight 328.77
Percent Purity 98.77%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.