missing translation for 'onlineSavingsMsg'
Learn More

Sulbutiamine, MedChemExpress

Product Code. 16492389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492389 1 g 1g
16431158 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492389 Supplier Medchem Express Supplier No. HYB2229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sulbutiamine is a synthetic analogue of vitamin B1 used for the treatment of asthenia.

Sulbutiamine is a synthetic analogue of vitamin B1 used for the treatment of asthenia.

Specifications

Chemical Name or Material Sulbutiamine
CAS 3286-46-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H46N8O6S2
Quantity 1 g
Synonym Bisibuthiamine
Solubility Information DMSO : ≥ 75 mg/mL (106.70 mM)
SMILES C/C(N(C=O)CC1=CN=C(C)N=C1N)=C(CCOC(C(C)C)=O)/SS/C(CCOC(C(C)C)=O)=C(C)\N(C=O)CC2=CN=C(C)N=C2N
Molecular Weight (g/mol) 702.89
Formula Weight 702.89
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.