missing translation for 'onlineSavingsMsg'
Learn More

Sulbactam, MedChemExpress

Product Code. 16408246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408246 Supplier Medchem Express Supplier No. HYB0334
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulbactam (CP45899) is a competitive, irreversible beta-lactamase inhibitor. Sulbactam shows antimicrobial activity against multidrug-resistant (MDR) acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex.

Sulbactam (CP45899) is a competitive, irreversible beta-lactamase inhibitor. Sulbactam shows antimicrobial activity against multidrug-resistant (MDR) acinetobacter calcoaceticus--Acinetobacter baumannii (Acb) complex.

Specifications

Chemical Name or Material Sulbactam
Physical Form Solid
CAS 68373-14-8
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H11NO5S
For Use With (Application) COVID-19-immunoregulation
Synonym CP45899
Solubility Information DMSO : ≥ 100 mg/mL (428.74 mM) ∣H2O : 20 mg/mL (85.75 mM; Need ultrasonic)
SMILES O=C([C@@H](C(C)(C)S([C@]1([H])C2)(=O)=O)N1C2=O)O
Molecular Weight (g/mol) 233.24
Formula Weight 233.24
Percent Purity 99.87%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.