missing translation for 'onlineSavingsMsg'
Learn More

Sulbactam, 98%

Product Code. 15477509
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477509 5 g 5g
15467509 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477509 Supplier Thermo Scientific Alfa Aesar Supplier No. J66235.06
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies. | CAS: 68373-14-8 | C8H11NO5S | 233.24 g/mol

Sulbactam is an antibacterial agent used in the treatment of multidrug resistant bacterium, Acinetobacter, a nosocomial pathogen. Also used in cell signaling studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68373-14-8
Molecular Formula C8H11NO5S
Molecular Weight (g/mol) 233.24
MDL Number MFCD00867005
InChI Key FKENQMMABCRJMK-RITPCOANSA-N
Synonym sulbactam, sulbactam acid, penicillanic acid sulfone, sulbactamum, betamaze, 2s,5r-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide, penicillanic acid 1,1-dioxide, unii-s4tf6i2330, penicillanic acid dioxide, chembl403
PubChem CID 130313
ChEBI CHEBI:9321
IUPAC Name (2S,5R)-3,3-dimethyl-4,4,7-trioxo-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

Specifications

Melting Point 154°C to 157°C
Packaging Glass bottle
Quantity 5 g
Solubility Information Soluble in water at 18mg/ml
Formula Weight 233.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulbactam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.