missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUHW1 Antibody (OTI2H3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74409AF700
This item is not returnable.
View return policy
Description
SUHW1 Monoclonal specifically detects SUHW1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| SUHW1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ZNF280A | |
| Recombinant protein fragment corresponding to amino acids 1-333 of human SUHW1 (NP_542778) produced in E.coli. | |
| 0.1 mL | |
| 129025.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI2H3 | |
| Western Blot, Flow Cytometry | |
| C19orf37, chromosome 19 open reading frame 37, Enzyme-like protein PIT13, MGC51082, Zfp428, zinc finger protein 428 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction