missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUGCT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555384
This item is not returnable.
View return policy
Description
Immunogen sequence: KGVKIIKELA AVCDVFVENY VPGKLSAMGL GYEDIDEIAP HIIYCSITGY GQTGPISQRA GYDAVASAVS GLMHITGPEN GDPVRPGVAM TDLATGLYAY GAIMAGLIQK YKTGKGLFID CNLLS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 29%.
SUGCT (Succinyl-CoA:Glutarate-CoA Transferase) is a Protein Coding gene. Diseases associated with SUGCT include Glutaric Aciduria Iii and Noonan Syndrome 8. Gene Ontology (GO) annotations related to this gene include succinate-hydroxymethylglutarate CoA-transferase activity. An important paralog of this gene is AMACR.
Specifications
| SUGCT | |
| Polyclonal | |
| Unconjugated | |
| Sugct | |
| 5033411D12Rik; C18H7orf10; C22orf6; C4H7orf10; C7orf10; D17907; dermal papilla derived protein 13; dermal papilla-derived protein 13; DERP13; GA3; hypothetical protein LOC619244; KIAA1186; KIAA1658; LOW QUALITY PROTEIN: succinate--hydroxymethylglutarate CoA-transferase; ORF19; RGD1308114; Russel-Silver syndrome candidate; succinate--hydroxymethylglutarate CoA-transferase; succinyl-CoA glutarate-CoA transferase; succinyl-CoA:glutarate-CoA transferase; succinylCoA:glutarate-CoA transferase; SUGCT; zgc:113390 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79783 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HAC7 | |
| Sugct | |
| Recombinant protein corresponding to Human SUGCT. Recombinant protein control fragment (Product #RP-92800). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction