missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Red 7B

Product Code. 10366691 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 10366691

Brand: Thermo Scientific Chemicals 191630050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6368-72-5 | C24H21N5 | 379.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6368-72-5
Molecular Formula C24H21N5
Molecular Weight (g/mol) 379.467
MDL Number MFCD00003904
InChI Key VKWNTWQXVLKCSG-UHFFFAOYSA-N
Synonym C.I. 26050, N-Ethyl-1-[4-(phenylazo)phenylazo]-2-naphthylamine, Fat Red 7B, Solvent Red 19
PubChem CID 61396
IUPAC Name N-ethyl-1-[(4-phenyldiazenylphenyl)diazenyl]naphthalen-2-amine
SMILES CCNC1=C(C2=CC=CC=C2C=C1)N=NC3=CC=C(C=C3)N=NC4=CC=CC=C4

Specifications

Melting Point 130.0°C
Beilstein 16, 379
Infrared Spectrum Authentic
Formula Weight 379.45
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Sudan red 7B
compliance-icons
Product Identifier
  • Sudan Red 7B
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.