missing translation for 'onlineSavingsMsg'
Learn More

Sudan R, TRC

Product Code. 30423372
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423372 1 g 1g
30421500 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30423372 Supplier Toronto Research Chemicals Supplier No. TRCS6889301G
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229-55-6
Molecular Formula C17 H14 N2 O2
Molecular Weight (g/mol) 278.31
Synonym 2-Naphthalenol, 1-[2-(2-methoxyphenyl)diazenyl]- (ACI), 1-[2-(2-Methoxyphenyl)diazenyl]-2-naphthalenol (ACI), 2-Naphthalenol, 1-[(2-methoxyphenyl)azo]- (9CI), 2-Naphthol, 1-(o-methoxyphenylazo)- (6CI), C.I. Solvent Red 1 (7CI, 8CI), Sudan Red G (6CI), 1-((2-Methoxyphenyl)diazenyl)naphthalen-2-ol, 1-(o-Anisylazo)-2-naphthol, 1-[(2-Methoxyphenyl)azo]-2-naphthol, Anisole-2-azo-β-naphthol, Brilliant Fat Scarlet R, C Red 2, C.I. 12150, C.I. Food Red 16, Ceres Red G, Ceres Red G 102, Fat Red BG, Fat Red G, Fat Red RS, Fat Soluble Red S, Food Red 16, Lacquer Red V 2G, NSC 11232, NSC 45194, Oil Pink, Oil Red, Oil Red 113, Oil Red OG, Oil Scarlet 389, Oil Soluble Red S, Oil Vermilion, Oil Vermilion LP, Oleal Red G, Organol Vermilion, Orient Oil Red OG, Plastoresin Red FR, Resinol Red G, Sico Fat Red BG New, Silotras Red TG, Solvent Red 1, Somalia Red PG, Sudan R, Sudan Red 290
IUPAC Name 1-[(2-methoxyphenyl)diazenyl]naphthalen-2-ol
SMILES COc1ccccc1N=Nc2c(O)ccc3ccccc23

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H14N2O2/c1-21-16-9-5-4-8-14(16)18-19-17-13-7-3-2-6-12(13)10-11-15(17)20/h2-11,20H,1H3
Formula Weight 278.1055
Chemical Name or Material Sudan Red G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.