missing translation for 'onlineSavingsMsg'
Learn More

Sudan Orange G, MP Biomedicals™

Product Code. 11495111 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495111 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495111 Supplier MP Biomedicals Supplier No. 0215670425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C.I. 11920, (Solvent Orange 1; 4-[Phenylazo]-resorcinol), Dye content: Approx. 95%

TRUSTED_SUSTAINABILITY

Specifications

CAS 2051-85-6
Molecular Formula C12H10N2O2
MDL Number MFCD00002275
InChI Key PULNESMFDLBKAZ-SDNWHVSQSA-N
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=C1
IUPAC Name (4E)-3-hydroxy-4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 214.22
PubChem CID 5921655
Formula Weight 214.2
Quantity 25 g
Physical Form Solid
Chemical Name or Material Sudan Orange G
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.