missing translation for 'onlineSavingsMsg'
Learn More

Sudan IV, Technical, C.I.26105, Fisher Chemical

Product Code. 11448862 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448862 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448862 Supplier Fisher Chemical Supplier No. S/8625/46
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 181.1°C
Boiling Point 260°C
CAS 85-83-6
Molecular Formula C24H20N4O
MDL Number MFCD00003893
InChI Key KMDLOETUWUPGMB-BXCCFQQFSA-N
SMILES CC1=CC(=CC=C1N\N=C1/C(=O)C=CC2=CC=CC=C12)N=NC1=CC=CC=C1C
IUPAC Name 1-[[2-methyl-4-[(2-methylphenyl)diazenyl]phenyl]hydrazinylidene]naphthalen-2-one
Molecular Weight (g/mol) 380.45
PubChem CID 6797604
Formula Weight 380.1662g/mol
Packaging Glass Jar
Color Brown/Red
Quantity 25 g
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • SUDAN IV
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.