missing translation for 'onlineSavingsMsg'
Learn More

Sudan III, MP Biomedicals™

Product Code. 11424411 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424411 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424411 Supplier MP Biomedicals Supplier No. 0215208780
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipophilic dye used as a biological stain for histological applications.

Specifications

CAS 85-86-9
Molecular Formula C22H16N4O
Synonym 1-[4-(phenylazo)phenylazo]-2-naphthol, Solvent Red 23
Color Brown/Red
Quantity 100 g
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Physical Form Powder
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
Molecular Weight (g/mol) 352.397
PubChem CID 6789251
Formula Weight 352.4
Grade Not Available
Chemical Name or Material Sudan III
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.