missing translation for 'onlineSavingsMsg'
Learn More

Sudan III, MP Biomedicals™

Product Code. 11414411 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414411 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414411 Supplier MP Biomedicals Supplier No. 0215208725
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used in coloring oils, sprit and lacquers. Sudan III, MP Biomedicals is a stain that is also used for zoological, pathological and vegetable objects (wax, cutin, resin, contents of lactiferous ducts, etc.) which are colored red, while cellulose membranes

TRUSTED_SUSTAINABILITY

Specifications

CAS 85-86-9
Melting Point 199°C (literature)
Molecular Formula C22H16N4O
Quantity 25 g
Synonym sudan iii, c.i. solvent red 23, 2-naphthalenol, 1-4-phenylazo phenyl azo, fhninjwbtrxebc-qszpnpogsa-n, sudaniii, sudan iiisolvent red 23, 2-naphthalenol, 1-2-4-2-phenyldiazenyl phenyl diazenyl, 1-2-4-2-phenyldiazen-1-yl phenyl diazen-1-yl naphthalen-2-ol, 1-e-4-e-phenylazo phenyl azo naphthalen-2-ol, 1-4-phenyldiazenylphenyl hydrazinylidene naphthalen-2-one
Solubility Information Insoluble in water; soluble in chloroform, glacial acetic acid; moderately soluble in alcohol, ether, acetone, fixed and volatile oils, hot glycerol. Clear red solution at 1mg/mL in toluence.
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
Molecular Weight (g/mol) 352.397
PubChem CID 6789251
Formula Weight 352.4
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.