missing translation for 'onlineSavingsMsg'
Learn More

Sudan III, MedChemExpress

Product Code. 16452429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452429 Supplier Medchem Express Supplier No. HYD0931
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sudan III is a hydrophobic bisazo dye.

Sudan III is a hydrophobic bisazo dye.

Specifications

Chemical Name or Material Sudan III
CAS 85-86-9
Purity Grade Notes Research
Molecular Formula C22H16N4O
Synonym Sudan Red III Tetrazobenzene-β-naphthol 111440 Red
Solubility Information DMSO : 2.27 mg/mL (6.44 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1=CC=C2C=CC=CC2=C1/N=N/C3=CC=C(/N=N/C4=CC=CC=C4)C=C3
Molecular Weight (g/mol) 352.39
Formula Weight 352.39
Grade Research
Color Reddish Brown
Quantity 1 g
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.