missing translation for 'onlineSavingsMsg'
Learn More

Sudan III (chromatographic standard), Thermo Scientific™

Product Code. p-7081468
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11410343

Brand: Thermo Scientific Alfa Aesar A11461.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
MDL Number MFCD00003905
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym C.I. 26100; Solvent Red 23
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Melting Point ∼200°C (decomposition)
Beilstein 931185
Merck Index 14,8884
Formula Weight 352.4
Quantity 25 g
Chemical Name or Material Sudan III (chromatographic standard)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.