missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan III

Product Code. 11493877
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493877 25 g 25g
11403887 100 g 100g
11413887 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493877 Supplier Thermo Scientific Chemicals Supplier No. A18318.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 85-86-9 | C22H16N4O | 352.397 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-86-9
Molecular Formula C22H16N4O
Molecular Weight (g/mol) 352.397
MDL Number MFCD00003905
InChI Key HTPQPMPFXUWUOT-UHFFFAOYSA-N
Synonym C.I. 26100; Solvent Red 23
PubChem CID 6789251
IUPAC Name 1-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalen-2-one
SMILES C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC=CC=C43

Specifications

Melting Point ∼200°C (decomposition)
Beilstein 931185
Merck Index 14,8884
Formula Weight 352.4
Quantity 25 g
Chemical Name or Material Sudan III

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.