missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan II, TRC
Specifications
Specifications
| Chemical Name or Material | Sudan II |
| CAS | 3118-97-6 |
| Molecular Formula | C18 H16 N2 O |
| Quantity | 5 g |
| Synonym | 2-Naphthalenol','1-[(2,4-dimethylphenyl)azo]- (9CI), C.I. Solvent Orange 7 (7CI,8CI), Sudan Red (6CI), 1-[2-(2,4-Dimethylphenyl)diazenyl]-2-naphthalenol, AF Red No. 5, Brasilazina Oil Scarlet 6G, Brilliant Oil Scarlet B, C.I. 12140, Calco Oil Scarlet BL, Ceres Orange RR, Cerisol Scarlet G, Ext D and C Red No. 14, FD and C Red No. 32, Fast Oil Orange II, Fat Scarlet 2G, Grasan Orange 3R, Japan Red 5, Japan Red 505, Japan Red No.5, Lacquer Orange VR, NSC 10457, Oil Orange 2R, Oil Orange KB, Oil Orange N Extra, Oil Orange R, Oil Orange X, Oil Orange XO, Oil Red XO, Oil Scarlet, Oil Scarlet 371, Oil Scarlet 6G, Oil Scarlet BL, Oil Scarlet YS, Red 505, Red No. 5, Resin Scarlet 2R, Solvent Orange 7, Somalia Orange 2R, Somalia Orange A 2R, Sudan 2, Sudan II, Sudan Orange RPA, Sudan Orange RRA, Sudan Red II, Sudan orange |
| InChI Formula | InChI=1S/C18H16N2O/c1-12-7-9-16(13(2)11-12)19-20-18-15-6-4-3-5-14(15)8-10-17(18)21/h3-11,21H,1-2H3 |
| SMILES | Cc1ccc(N=Nc2c(O)ccc3ccccc23)c(C)c1 |
| IUPAC Name | 1-[(E)-(2,4-dimethylphenyl)diazenyl]naphthalen-2-ol |
| Molecular Weight (g/mol) | 276.33 |
| Formula Weight | 276.1263 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?