missing translation for 'onlineSavingsMsg'
Learn More

Sudan II, TRC

Product Code. 30424256
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424256 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424256 Supplier Toronto Research Chemicals Supplier No. TRCS688890500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sudan II
CAS 3118-97-6
Molecular Formula C18 H16 N2 O
Quantity 500 mg
Synonym 2-Naphthalenol','1-[(2,4-dimethylphenyl)azo]- (9CI), C.I. Solvent Orange 7 (7CI,8CI), Sudan Red (6CI), 1-[2-(2,4-Dimethylphenyl)diazenyl]-2-naphthalenol, AF Red No. 5, Brasilazina Oil Scarlet 6G, Brilliant Oil Scarlet B, C.I. 12140, Calco Oil Scarlet BL, Ceres Orange RR, Cerisol Scarlet G, Ext D and C Red No. 14, FD and C Red No. 32, Fast Oil Orange II, Fat Scarlet 2G, Grasan Orange 3R, Japan Red 5, Japan Red 505, Japan Red No.5, Lacquer Orange VR, NSC 10457, Oil Orange 2R, Oil Orange KB, Oil Orange N Extra, Oil Orange R, Oil Orange X, Oil Orange XO, Oil Red XO, Oil Scarlet, Oil Scarlet 371, Oil Scarlet 6G, Oil Scarlet BL, Oil Scarlet YS, Red 505, Red No. 5, Resin Scarlet 2R, Solvent Orange 7, Somalia Orange 2R, Somalia Orange A 2R, Sudan 2, Sudan II, Sudan Orange RPA, Sudan Orange RRA, Sudan Red II, Sudan orange
InChI Formula InChI=1S/C18H16N2O/c1-12-7-9-16(13(2)11-12)19-20-18-15-6-4-3-5-14(15)8-10-17(18)21/h3-11,21H,1-2H3
SMILES Cc1ccc(N=Nc2c(O)ccc3ccccc23)c(C)c1
IUPAC Name 1-[(E)-(2,4-dimethylphenyl)diazenyl]naphthalen-2-ol
Molecular Weight (g/mol) 276.33
Formula Weight 276.1263
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.