missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan II, pure

Product Code. 10328330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10328330 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10328330 Supplier Thermo Scientific Chemicals Supplier No. 190150250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3118-97-6 | C18H16N2O | 276.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 156.0°C to 158.0°C
CAS 3118-97-6
Molecular Formula C18H16N2O
MDL Number MFCD00003896
Beilstein 16, 168
Synonym C.I. 12140, 1-(2, 4-Dimethylphenylazo)-2-naphthol, Solvent orange 7
InChI Key YPXOPAFVVHXQDP-ZZEZOPTASA-N
Infrared Spectrum Authentic
SMILES CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1
IUPAC Name (1Z)-1-[(2,4-dimethylphenyl)hydrazinylidene]naphthalen-2-one
Molecular Weight (g/mol) 276.34
PubChem CID 5809936
Lambda Max at approximately 420 and 493nm (in methanol)
Formula Weight 276.33
Grade Pure
Packaging Glass bottle
Specific Extinction 500 min. (at 493nm, in methanol)
Color Brown/Orange to Red
Quantity 25 g
Physical Form Powder
Chemical Name or Material Sudan II
Show More Show Less
compliance-icons
Product Identifier
  • Sudan II
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.