missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan II

Product Code. 11325119
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11325119 100 g 100g
11315119 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325119 Supplier Thermo Scientific Chemicals Supplier No. A17613.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3118-97-6 | C18H16N2O | 276.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3118-97-6
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.34
MDL Number MFCD00003896
InChI Key YPXOPAFVVHXQDP-ZZEZOPTASA-N
Synonym C.I. 12140; Solvent Orange 7
PubChem CID 5809936
IUPAC Name (1Z)-1-[2-(2,4-dimethylphenyl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES CC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(C)=C1

Specifications

Melting Point 156°C to 160°C
Formula Weight 276.34
Quantity 100 g
Chemical Name or Material Sudan II

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.