missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sudan I, TRC
Specifications
Specifications
| Chemical Name or Material | Sudan I |
| CAS | 842-07-9 |
| Molecular Formula | C16 H12 N2 O |
| Quantity | 100 mg |
| Synonym | 2-Naphthalenol','1-(phenylazo)- (9CI), 2-Naphthol','1-phenylazo- (3CI), C.I. Solvent Yellow 14 (8CI), Sudan Yellow (6CI), 1-(2-Phenyldiazenyl)-2-naphthalenol, 1-(2-Hydroxynaphthyl)azobenzene, 1-(Phenylazo)-2-hydroxynaphthalene, 1-(Phenylazo)-2-naphthol, 1-Benzeneazo-2-naphthol, 1-Phenylazo-2-naphthalenol, 1-Phenylazo-β-naphthol, 2-Hydroxy-1-(phenylazo)naphthalene, 2-Hydroxynaphthyl-1-azobenzene, Benzeneazo-β-naphthol, Brasilazina Oil Orange, C.I. 12055, C.I. Disperse Yellow 97, Calcogas Orange NC, Ceres Orange R, Disperse Yellow 97, Dispersol Fast Orange B, Dispersol Yellow PP, Enial Orange I, Fast Oil Orange I, Fast Orange, Fat Orange 4A, Fat Orange G, Fat Orange I, Fat Orange R, Fat Orange R 01, Fat Orange RS, Fat Soluble Orange, Grasal Orange, Grasan Orange R, Hidaco Oil Orange, Lacquer Orange VG, Morton Orange Y, NSC 11227, NSC 51524, Oil Orange, Oil Orange 2311, Oil Orange 2B, Oil Orange 31, Oil Orange 5101, Oil Orange 7078-V, Oil Orange E, Oil Orange PEL, Oil Orange R, Oil Orange Z-7078, Oil Soluble Orange, Oil Soluble Yellow R, Oil orange PS, Oleal Orange R, Orange 3RA Soluble in Grease, Orange JLS-PS, Orange Pel, Orange R Fat Soluble, Organol Orange, Orient Oil Orange PS, Petrol Orange Y, Plastoresin Orange F 4A, Resinol Orange R, Silotras Orange TR, Solvent Yellow 14, Somalia Orange I, Stearix Orange, Sudan 1, Sudan I, Sudan Orange 220, Sudan Orange R, Sudan Orange RA, Sudan Orange RA New, Sudan Red I, Tertrogras Orange SV, Toyo Oil Orange, Waxoline Orange EP-FW, Waxoline Yellow I, Waxoline Yellow IM, Waxoline Yellow IP, Waxoline Yellow IS, α-Phenylazo-β-naphthol |
| InChI Formula | InChI=1S/C16H12N2O/c19-15-11-10-12-6-4-5-9-14(12)16(15)18-17-13-7-2-1-3-8-13/h1-11,19H |
| SMILES | Oc1ccc2ccccc2c1N=Nc3ccccc3 |
| IUPAC Name | 1-[(E)-phenyldiazenyl]naphthalen-2-ol |
| Molecular Weight (g/mol) | 248.28 |
| Formula Weight | 248.095 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?