missing translation for 'onlineSavingsMsg'
Få mere at vide

MedChemExpress Sudan I

Artikelnummer. 16412409
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 g
Pakningsstørrelse:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16412409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16412409 Leverandør MedChemExpress Leverandørnr. HYD0024

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Sudan I (Solvent Yellow 14) is a diazo-conjugate red dye and can be used as an additive to products such as oils, solvents or polishes. Sudan I inhibits growth of bacterial strains Clostridium perfringens and L. rhamnosus.

Tekniske data

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
CAS 842-07-9
Molecular Weight (g/mol) 248.28 g/mol
Synonym Solvent Yellow 14
Target HBV
Solubility DMSO : 50 mg/mL (201.39 mM; Need ultrasonic) ∣ H2O : 0.67 mg/mL (2.70 mM; Need ultrasonic)
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.