missing translation for 'onlineSavingsMsg'
Learn More

Sudan Blue II;Oil Blue 35, TRC

Product Code. 30421111
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421111 5 g 5g
30427544 500 mg 500mg
30412083 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30421111 Supplier Toronto Research Chemicals Supplier No. TRCS6889135G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17354-14-2
Molecular Formula C22 H26 N2 O2
Molecular Weight (g/mol) 350.45
Synonym 9,10-Anthracenedione, 1,4-bis(butylamino)- (9CI, ACI), 1,4-Bis(butylamino)-9,10-anthracenedione (ACI), Anthraquinone, 1,4-bis(butylamino)- (7CI, 8CI), 1,4-Bis(butylamino)-9,10-anthraquinone, 1,4-Bis(butylamino)anthraquinone, Aizen SOT Blue 2, Blue 2N, C Ext. Blue 12, C.I. 61554, C.I. Solvent Blue 35, Fat Blue B, Fat Blue B 01, Hytherm Blue B 200, Kayaset Blue N, KP Plast Blue BR, Oil Blue 2N, Oil Blue 35, Oil Blue 5502, Oil Blue B, Oil Soluble Blue 11, Oplas Blue 2N, Oplas Blue IIn, Opurous Blue IIN, Orcosolve Blue B, Orient Oil Blue 2N, Orimax Blue 2N, Sico Fat Blue 50401N, Solvent Blue 35, SOT-Blue 2, Sudan Blue 2, Sudan Blue 670, Sudan Blue II, Sudan Blue II Base, Sumiplast Blue S
IUPAC Name 1,4-bis(butylamino)anthracene-9,10-dione
SMILES CCCCNc1ccc(NCCCC)c2C(=O)c3ccccc3C(=O)c12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H26N2O2/c1-3-5-13-23-17-11-12-18(24-14-6-4-2)20-19(17)21(25)15-9-7-8-10-16(15)22(20)26/h7-12,23-24H,3-6,13-14H2,1-2H3
Formula Weight 350.1994
Chemical Name or Material Sudan Blue II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.