missing translation for 'onlineSavingsMsg'
Learn More

Sudan Black, TRC

Product Code. 30138669
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138669 5 g 5g
30137823 25 g 25g
17933375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138669 Supplier Toronto Research Chemicals Supplier No. TRCS6889105G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29 H24 N6
Molecular Weight (g/mol) 456.54
Synonym 1H-Perimidine, 2,3-dihydro-2,2-dimethyl-6-[2-[4-(2-phenyldiazenyl)-1-naphthalenyl]diazenyl]- (ACI), 1H-Perimidine, 2,3-dihydro-2,2-dimethyl-6-[[4-(phenylazo)-1-naphthalenyl]azo]- (9CI), 2,3-Dihydro-2,2-dimethyl-6-[2-[4-(2-phenyldiazenyl)-1-naphthalenyl]diazenyl]-1H-perimidine (ACI), C.I. Solvent Black 3 (7CI, 8CI), Sudan Black B (6CI), (2,2-Dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene, 2,2-Dimethyl-6-[[4-(phenyldiazenyl)-1-naphthyl]diazenyl]-2,3-dihydro-1H-perimidine, 2,3-Dihydro-2,2-dimethyl-6-[(4-phenylazo-1-naphthalenyl)-azo]-1H-perimidine, 2,3-Dihydro-2,2-dimethyl-6-[[4-(phenylazo)-1-naphthyl]azo]perimidine, 6-(1-Phenylazo-4-naphthylazo)-2,3-dihydro-2,2-dimethylperimidine, Aizen SOT Black 6, C.I. 26150, Ceres Black BN, Chuo Sudan Black 141, Fast Black HBN, Fat Black HB, Fat Black HB 01, Hexatype Black B, Lacquer Black S, Lacquer Black VB, Neptune Black X 60, NSC 11239, Nubian Black BT, Oil Black 141, Oil Black 860, Oil Black BT, Oil Black HBB, Oil Black HZ, Orient Oil Black 860, Orient Oil Black HBB, Polysolve Black 3, Solvent Black 3, SOT-Black 6, Sudan Black, Sudan Black 141, Sudan Black BB, Sudan Black X 60, Sudan Deep Black BB, Sudan Deep Black BN, Typogen Black
IUPAC Name (E)-(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-[4-[(E)-phenyldiazenyl]naphthalen-1-yl]diazene
SMILES CC1(C)Nc2cccc3c(ccc(N1)c23)N=Nc4ccc(N=Nc5ccccc5)c6ccccc46

Specifications

Quantity 5 g
InChI Formula InChI=1S/C29H24N6/c1-29(2)30-26-14-8-13-22-25(17-18-27(31-29)28(22)26)35-34-24-16-15-23(20-11-6-7-12-21(20)24)33-32-19-9-4-3-5-10-19/h3-18,30-31H,1-2H3
Formula Weight 456.2062
Chemical Name or Material Sudan Black B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.