missing translation for 'onlineSavingsMsg'
Learn More

Sudan Black B, MP Biomedicals

Product Code. 11436895 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436895 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436895 Supplier MP Biomedicals Supplier No. 0520647010
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4197-25-5
Molecular Formula C29H24N6
Quantity 10 g
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
Molecular Weight (g/mol) 456.553
PubChem CID 61336
Formula Weight 456.55
Chemical Name or Material Sudan Black B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.