missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Black B, high purity biological stain

Product Code. 10475125 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10475125

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4197-25-5 | C29H24N6 | 456.553 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29H24N6
Molecular Weight (g/mol) 456.553
MDL Number MFCD00006919
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
Synonym C.I. 26150, Fat Black HB, Solvent Black 3
PubChem CID 61336
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C

Specifications

Melting Point 120.0°C to 124.0°C
Solubility Information Solubility in water: insoluble.
Infrared Spectrum Authentic
Formula Weight 456.54
Grade High Purity
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Brown
Quantity 100 g
Physical Form Fine Crystalline Powder
Chemical Name or Material Sudan Black B, high purity biological stain
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.