missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Suction Tube Transport Case

Product Code. 11802293 Shop All Bürkle Products
Change view
Click to view available options
Color:
Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11802293 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11802293 Supplier Buerkle™ Supplier No. 53051029
  • ONLINE EXCLUSIVE PRICE
    £158.00
    £148.72 / Pack of 1
    Save £9.28
    5% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the practical transport of new suction tubes and the safe storage of used ones, suitable for 100 suction tubes, 220 cm long

  • For the practical transport of unused suction tubes and for safe storage of used ones
  • Accessory part for the ProfiSampler sampler
TRUSTED_SUSTAINABILITY

Specifications

Product Type Transport Case
Color Gray
For Use With (Equipment) For Straight Suction Tubes
Weight (Metric) 0.8 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.