Learn More
Suction Syphon Pump with Polyethylene Suction Tube
Product Code. 12997954
Encompass_Preferred
Change view
Click to view available options
Up to 25L Drums
Product Code.
12997954 Up to 25L Drums
1 options
Product Code. 12997954 Supplier Brand Not Defined Supplier No. 5915
Only null left
Add to basket
Add to basket
A selection of polyethylene pumps. They can be used either as a pump or in syphoning mode. For safely dispensing liquids from containers ranging from 5L up to 205L drums
Specifications
Quantity 1 Each
Type Suction Syphon Pump
For Use With (Application) For safely dispensing liquids from containers ranging from 25L drums