missing translation for 'onlineSavingsMsg'
Learn More

Sucrose,Ultra Pure, RNase Free, 1kg, MP Biomedicals

Product Code. 11431732 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431732 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11431732 Supplier MP Biomedicals Supplier No. 04821713
  • ONLINE EXCLUSIVE PRICE
    £117.83
    £102.59 / 1kg
    Save £15.24
    12% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultra-pure, RNase-free sucrose powder suitable for use in density gradients.

Specifications

CAS 57-50-1
Molecular Formula C12H22O11
MDL Number MFCD00006626
Quantity 1 kg
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 342.30
PubChem CID 5988
ChEBI CHEBI:17992
Grade Ultrapure
Physical Form Crystalline Powder
Chemical Name or Material Sucrose, Ultra Pure, RNase Free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.