missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sucrose stearate

Product Code. 15438369
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438369 25 g 25g
15428369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438369 Supplier Thermo Scientific Chemicals Supplier No. J66845.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25168-73-4 | C30H56O12 | 608.766 g/mol

Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sucrose stearate is used as an emulsifier in many beauty applications and cosmetics such as makeup products, cleansing formulas. Emulsifiers aid the blending of water and fatty substances, a common ingredient in many foods.

Solubility
Soluble in water and ethanol at 75°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25168-73-4
Molecular Formula C30H56O12
Molecular Weight (g/mol) 608.766
MDL Number MFCD00152822
InChI Key SZYSLWCAWVWFLT-UTGHZIEOSA-N
Synonym sucrose stearate, sucrose, 1-stearate, unii-58rp7ju52k, sucrose stearate ester, alpha-d-glucopyranoside, 1-o-1-oxooctadecyl-beta-d-fructofuranosyl, 1-o-stearoyl-beta-d-fructofuranosyl alpha-d-glucopyranoside, unii-274kw0o50m component, unii-d0951ov2o1 component, unii-l98x941w2b component, 2s,3s,4s,5r-3,4-dihydroxy-5-hydroxymethyl-2-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxolan-2-yl methyl octadecanoate
PubChem CID 9898327
IUPAC Name [(2S,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-2-yl]methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC1(C(C(C(O1)CO)O)O)OC2C(C(C(C(O2)CO)O)O)O

Specifications

Color White
Quantity 25 g
Solubility Information Soluble in water and ethanol at 75°C.
Formula Weight 608.75
Physical Form Powder
Chemical Name or Material Sucrose stearate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.