missing translation for 'onlineSavingsMsg'
Learn More

Sucrose octaacetate, 98%

Product Code. 11996901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
Unit Size:
1kg
250g
5g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11996901 5 kg Plastic drum 5kg
10742792 5 g Glass Bottle 5g
10752792 250 g Plastic Bottle 250g
10677571 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11996901 Supplier Thermo Scientific Acros Supplier No. 419790051
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 126-14-7 | C28H38O19 | 678.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-14-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.59
InChI Key ZIJKGAXBCRWEOL-SAXBRCJISA-N
Synonym sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate
PubChem CID 31340
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point >78.0°C
Color Cream-White to White
Density 1.2800g/mL
Boiling Point 250.0°C (1.0 mmHg)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Quantity 5 kg
Merck Index 15,9013
Specific Gravity 1.28
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetic acid,acetone,benzene,90g/l in ethanol
Formula Weight 678.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sucrose octaacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.