missing translation for 'onlineSavingsMsg'
Learn More

Sucrose Monolaurate, TRC

Product Code. 30414964
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414964 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414964 Supplier Toronto Research Chemicals Supplier No. TRCS080898500MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sucrose Monolaurate
CAS 25339-99-5
Molecular Formula C24H44O12
Quantity 500 mg
Synonym Cane Sugar Monolaurate, D 1216, DK Ester L 160, DK Ester S-L 18A, LWA 1570, Nitto Ester LW 1540, Nitto LW 1540, Ryoto LWA 1570, Ryoto Sugar Ester LWA 1540, Ryoto Sugar Ester LWA 1570, Ryoto Sugar Ester MO-C 12, Ryoto Sugar Ester SM 1200, SM 1200, Saccharose Laurate, Sucrose Lauric Acid Ester, Sucrose Lauric Acid Monoester, Sucrose Lauryl Ester, Sucrose Monododecanoate, Sucrose Monolaurate, Sucrose Monolauryl Ester, Surfhope SE Cosme C 1216, Surfhope SE Pharma D 1216, Dodecanoyl D-Sucrose
InChI Formula InChI=1S/C24H44O12/c1-2-3-4-5-6-7-8-9-10-11-17(27)33-13-16-18(28)20(30)21(31)23(34-16)36-24(14-26)22(32)19(29)15(12-25)35-24/h15-16,18-23,25-26,28-32H,2-14H2,1H3/t15-,16-,18-,19-,20+,21-,22+,23-,24+/m1/s1
SMILES O[C@H]1[C@H](O)[C@@](O[C@@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](COC(CCCCCCCCCCC)=O)O2)(CO)O[C@@H]1CO
IUPAC Name ((2R,3S,4S,5R,6R)-6-(((2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)methyl dodecanoate
Molecular Weight (g/mol) 524.6
Formula Weight 524.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.