missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, MedChemExpress
Description
Sucrose (D-(+)-Saccharose) is a disaccharide which is composed of two monosaccharides, glucose and fructose. Sucrose can be applied in some animal models, including metabolic disease, obesity, diet on preference, and diabetes, et al.
Specifications
Specifications
| Chemical Name or Material | Sucrose |
| CAS | 57-50-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H22O11 |
| Quantity | 100 mg |
| Synonym | D-(+)-Saccharose |
| Solubility Information | H2O : 100 mg/mL (292.14 mM; Need ultrasonic and warming) ∣DMSO : 100 mg/mL (292.14 mM; Need ultrasonic) |
| SMILES | O[C@H]1[C@H](O)[C@@H](CO)O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@@H]1O |
| Molecular Weight (g/mol) | 342.3 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?