missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, for Electrophoresis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
3 kg
Unit Size:
3kg
Specifications
Specifications
| Melting Point | 190°C |
| Color | White |
| pH | 6.5 to 7.5 |
| Packaging | Plastic powder jar |
| Quantity | 3 kg |
| Formula Weight | 342.29g/mol |
| Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction