missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, (Crystalline), MP Biomedicals

Product Code. 11435702 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
Unit Size:
1lb
25lb
5lb
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435702 25 lb. 25lb
11415702 1 lb. 1lb
11425702 5 lb. 5lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435702 Supplier MP Biomedicals Supplier No. 0290297825
  • ONLINE EXCLUSIVE PRICE
    £179.00
    £166.95 / 25lb
    Save £12.05
    6% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This is a slightly less pure sucrose than our 190271. It does not reduce Fehling's solution, and is suitable for most routine applications

  • Its a simple carbohydrate.
  • It is used in density gradients, Dielectrophoretic Field-Flow Fractionation running buffer.
  • It is a main component of media and animal diet.
  • Sucrose is a simple carbohydrate, a disaccharide composed of glucose and fructose.
  • It is hydrolyzed to glucose and fructose by dilute acids and by invertase, a yeast enzyme

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
PubChem CID 5988
ChEBI CHEBI:17992
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Quantity 25 lb.
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Formula Weight 342.3
Grade Food
Physical Form Crystalline
Chemical Name or Material Sucrose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.