missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, ACS, Thermo Scientific Chemicals
Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
Specifications
Specifications
| Melting Point | 185°C to 187°C |
| Density | 1.58 |
| Packaging | Plastic bottle |
| Quantity | 500 g |
| Beilstein | 90825 |
| Specific Gravity | 1.58 |
| Formula Weight | 342.3 |
| Grade | ACS |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sucrose |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction