missing translation for 'onlineSavingsMsg'
Learn More

Sucralose, Thermo Scientific™

Product Code. p-7227392 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
This item is not returnable. View return policy

Product Code. 17740689

Brand: Thermo Scientific Acros 470091000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Alpha Vector SUCRALOSE
Chemical Name or Material Sucralose
CAS 56038-13-2
Melting Point 130°C
Color White
Infrared Spectrum Conforms
Packaging Plastic bottle
Molecular Formula C12 H19 Cl3 O8
MDL Number 03648615
Quantity 100 g
Merck Index XIII 8965
Specific Rotation Condition (20°C, 589 nm) (c=1% aq. soln.)
Specific Rotation 84° to 88°
Synonym Trichlorosucrose
Molecular Weight (g/mol) 397.63
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.