missing translation for 'onlineSavingsMsg'
Learn More

Sucralose, MedChemExpress

Product Code. 16442386
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442386 500 mg 500mg
16439116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442386 Supplier Medchem Express Supplier No. HYN0614
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sucralose (E955; Trichlorosucrose) is a non-nutritive artificial sweetener and sugar substitute. Sucralose can activate a conserved neural fasting response and thereby exerts an appetite-stimulating effect in rodents.

Sucralose (E955

Specifications

Chemical Name or Material Sucralose
CAS 56038-13-2
Color White
Purity Grade Notes Research
Molecular Formula C12H19Cl3O8
Quantity 500 mg
Synonym E955 Trichlorosucrose
Solubility Information H2O : 100 mg/mL (251.49 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (251.49 mM; Need ultrasonic)
SMILES ClC[C@]1(O[C@H](CCl)[C@@H](O)[C@@H]1O)O[C@H]([C@@H]([C@@H](O)[C@H]2Cl)O)O[C@@H]2CO
Molecular Weight (g/mol) 397.63
Formula Weight 397.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.