missing translation for 'onlineSavingsMsg'
Learn More

Sucralfate, MedChemExpress

Product Code. 16482186
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16482186

Brand: Medchem Express HYB0644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sucralfate (Sucrose octasulfate-aluminum complex) is a potent and orally active gastroprotectant with no systemic effects. Sucralfate inhibits peptic activity and binds to negatively charged subepithelial proteins exposed during mucosal injury, forming a viscous layer that protects the vascular bed and proliferative zone. Sucralfate is used for prevention and treatment of several gastrointestinal diseases in vivo.

Sucralfate (Sucrose octasulfate-aluminum complex) is a potent and orally active gastroprotectant with no systemic effects. Sucralfate inhibits peptic activity and binds to negatively charged subepithelial proteins exposed during mucosal injury, forming a viscous layer that protects the vascular bed and proliferative zone. Sucralfate is used for prevention and treatment of several gastrointestinal diseases in vivo.

Specifications

Chemical Name or Material Sucralfate
Physical Form Powder
CAS 54182-58-0
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H14Al16O75S8
For Use With (Application) COVID-19-immunoregulation
Synonym Sucrose octasulfate-aluminum complex
Solubility Information DMSO : < 1 mg/mL (insoluble or slightly soluble) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [OSO3[Al2(OH5)]][C@H]([C@@H](C[OSO3[Al2(OH5)]])O1)[C@H](O[S]=O=O=O=[OH5]#[Al]#[Al])[C@]1(C[OSO3[Al2(OH5)]])O[C@@H]2[C@H](O[S]=O=O=O=[OH5]#[Al]#[Al])[C@H](O[S]=O=O=O=[OH5]#[Al]#[Al])[C@H](O[S]=O=O=O=[OH5]#[Al]#[Al])[C@@H](C[OSO3[Al2(OH5)]])O2
Molecular Weight (g/mol) 2046.42
Formula Weight 2046.42
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.