missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SUCLG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557766
This item is not returnable.
View return policy
Description
Immunogen sequence: LPQNGIRHCS YTASRQHLYV DKNTKIICQG FTGKQGTFHS QQALEYGTKL VGGTTPGKGG QTHLGLPVFN TVKEAKEQTG A Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
This gene encodes the alpha subunit of the heterodimeric enzyme succinate coenzyme A ligase. This enzyme is targeted to the mitochondria and catalyzes the conversion of succinyl CoA and ADP or GDP to succinate and ATP or GTP. Mutations in this gene are the cause of the metabolic disorder fatal infantile lactic acidosis and mitochondrial DNA depletion.
Specifications
| SUCLG1 | |
| Polyclonal | |
| Unconjugated | |
| SUCLG1 | |
| 1500000I01Rik; GALPHA; MTDPS9; SCS-alpha; Succinate--CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinate-CoA ligase alpha subunit; succinate-CoA ligase, alpha subunit; succinate-CoA ligase, GDP-forming alpha subunit; succinate-CoA ligase, GDP-forming, alpha subunit; succinyl-CoA ligase [ADP/GDP-forming] subunit alpha, mitochondrial; succinyl-CoA ligase [GDP-forming] subunit alpha, mitochondrial; succinyl-CoA synthetase alpha subunit; succinyl-CoA synthetase subunit alpha; SUCLA1; Suclg1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114597, 56451, 8802 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P13086, P53597, Q9WUM5 | |
| SUCLG1 | |
| Recombinant protein corresponding to Human SUCLG1. Recombinant protein control fragment (Product #RP-96187). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction